missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SHMT1 Antibody, DyLight 680, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-32173FR
This item is not returnable.
View return policy
Description
SHMT1 Polyclonal antibody specifically detects SHMT1 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| SHMT1 | |
| Polyclonal | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| 14 kDa protein, CSHMT, cytoplasmic serine hydroxymethyltransferase, Glycine hydroxymethyltransferase, MGC15229, MGC24556, serine hydroxymethyltransferase 1 (soluble), serine hydroxymethyltransferase, cytosolic, Serine methylase, SHMTEC 2.1.2.1 | |
| The immunogen this antibody was made to, maps to a region between residue 453 to 483 of human Serine Hydroxymethyltransferase 1 using the numbering given in entry NP_004160.3 (GeneID 6470). | |
| 0.1 mL | |
| metabolism | |
| 6470 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| DyLight 680 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction