missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SHMT2 Monoclonal Antibody (OTI1C1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808839
This item is not returnable.
View return policy
Description
Mammalian serine hydroxymethyltransferase (SHMT) is a tetrameric, pyridoxal phosphate (PLP)-dependent enzyme that catalyzes the reversible interconversion of serine and tetrahydrofolate to glycine and methylenetetrahydrofolate in the cytoplasm (cSHMT, SHMT1) and mitochondria (mSHMT, SHMT2). cSHMT preferentially supplies one-carbon units for thymidylate biosynthesis, depletes methylenetetrahydrofolate pools for S-adenosylmethionine (SAM) synthesis by synthesizing serine, sequesters 5-methyltetrahydrofolate, and inhibits SAM synthesis.Specifications
| SHMT2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P34897 | |
| SHMT2 | |
| Full length human recombit protein of human SHMT2 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Western Blot | |
| OTI1C1 | |
| Unconjugated | |
| SHMT2 | |
| GLYA, HEL-S-51e, SHMT | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 6472 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction