Learn More
SHMT2 Mouse anti-Human, Clone: OTI1C1, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF808839
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Mammalian serine hydroxymethyltransferase (SHMT) is a tetrameric, pyridoxal phosphate (PLP)-dependent enzyme that catalyzes the reversible interconversion of serine and tetrahydrofolate to glycine and methylenetetrahydrofolate in the cytoplasm (cSHMT, SHMT1) and mitochondria (mSHMT, SHMT2). cSHMT preferentially supplies one-carbon units for thymidylate biosynthesis, depletes methylenetetrahydrofolate pools for S-adenosylmethionine (SAM) synthesis by synthesizing serine, sequesters 5-methyltetrahydrofolate, and inhibits SAM synthesis.Specifications
| SHMT2 | |
| Monoclonal | |
| Unconjugated | |
| SHMT2 | |
| GLYA, HEL-S-51e, SHMT | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 6472 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI1C1 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| P34897 | |
| SHMT2 | |
| Full length human recombit protein of human SHMT2 produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.