Learn More
Short-Length Microcaps™ Micropipets
Ideal for handling dangerous materials
Brand: N/A
Additional Details : Weight : 0.50000kg
Includes: Bulb assembly
Description
Precision-made capillary glass tubing calibrated “To Contain” definite volume from end to end. Accurate to within ±1%.
- Capacity: 5μL
- To use, insert pipet through cap on end of rubber bulb assembly until visible through glass tube
- Hole in rubber bulb permits free air circulation, fills pipet by capillary action
- Pipet contains stated volume when completely filled from end to end
- To discharge, hold finger over hole in bulb and squeeze
- To rinse, repeat operation with diluent
Order Info
Packed in vials of 100.
†Heparinized.
Specifications
±1% | |
Bulb assembly | |
Microcaps™ Micropipet |
Glass | |
5 μL |