Learn More
Short-Length Microcaps™ Micropipets
Ideal for handling dangerous materials
Brand:  N/A
Additional Details : Weight : 0.50000kg
Product Code. 10134344
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Includes: Bulb assembly
Description
Description
Precision-made capillary glass tubing calibrated “To Contain” definite volume from end to end. Accurate to within ±1%.
  • Capacity: 5μL
  • To use, insert pipet through cap on end of rubber bulb assembly until visible through glass tube
  • Hole in rubber bulb permits free air circulation, fills pipet by capillary action
  • Pipet contains stated volume when completely filled from end to end
  • To discharge, hold finger over hole in bulb and squeeze
  • To rinse, repeat operation with diluent
Order Info
Packed in vials of 100.
†Heparinized.
Specifications
Specifications
±1%
Bulb assembly
Microcaps™ Micropipet
Glass
5 μL