missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ SHP-1 Antibody (PTPN6/7543) [Alexa Fluor™ 532], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-26999AF532
This item is not returnable.
View return policy
Description
SHP-1 Monoclonal antibody specifically detects SHP-1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| SHP-1 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| HCP, HCPH, HPTP1C, PTP-1C, PTPN6, SHP1, SHP-1L, SH-PTP1 | |
| Recombinant fragment (around aa395-595) of human SHP-1 protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Apoptosis, Phospho Specific, Protein Phosphatase | |
| 5777 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Immunohistochemistry (Paraffin) | |
| PTPN6/7543 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction