missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SHP-2/PTPN11 Antibody (M163), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-23400
This item is not returnable.
View return policy
Description
SHP-2/PTPN11 Monoclonal antibody specifically detects SHP-2/PTPN11 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| SHP-2/PTPN11 | |
| Monoclonal | |
| Unconjugated | |
| PBS, 1 mg/ml BSA and 50% glycerol | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| M163 | |
| Western Blot 1:1000 | |
| BPTP3, EC 3.1.3.48, MGC14433, protein tyrosine phosphatase, non-receptor type 11, protein tyrosine phosphatase-2, Protein-tyrosine phosphatase 1D, Protein-tyrosine phosphatase 2C, PTP-1D, PTP-2C, PTP2CNS1, Shp2, SHP-2, SHP2CFC, SHPTP2, SH-PTP2Noonan syndrome 1, SH-PTP3, tyrosine-protein phosphatase non-receptor type 11 | |
| Clone (M163) was generated from a recombinant protein containing amino acids in the N-terminal region of human SHP-2/PTPN11. This sequence is highly conserved in rat and mouse SHP-2/PTPN11. [UniProt# Q06124] | |
| 0.1 mL | |
| Angiogenesis, Cancer, Cell Biology, Lipid and Metabolism, Phospho Specific, Signal Transduction, Tyrosine Kinases | |
| 5781 | |
| Store at -20°C. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction