Learn More
SHP-2/PTPN11 Antibody (OTI3F2), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-74173AF750
Product Code. 18164317
  • € 785.00 / 0.10mL
Description
SHP-2/PTPN11 Monoclonal specifically detects SHP-2/PTPN11 in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SHP-2/PTPN11
Monoclonal
Alexa Fluor 750
50 mM sodium borate with 0.05% sodium azide
PTPN11
Full length recombinant protein of human SHP-2/PTPN11 (NP_002825) produced in HEK293T cells.
Primary
Human, Mouse, Rat, Canine
IgG2a
Western Blot, Flow Cytometry
OTI3F2
Western Blot, Flow Cytometry
BPTP3, EC 3.1.3.48, MGC14433, protein tyrosine phosphatase, non-receptor type 11, protein tyrosine phosphatase-2, Protein-tyrosine phosphatase 1D, Protein-tyrosine phosphatase 2C, PTP-1D, PTP-2C, PTP2CNS1, Shp2, SHP-2, SHP2CFC, SHPTP2, SH-PTP2Noonan syndrome 1, SH-PTP3, tyrosine-protein phosphatase non-receptor type 11
Mouse
0.1 mL
5781
Store at 4°C in the dark.