Learn More
SHPK Monoclonal Antibody (OTI1D2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA503209S
This item is not returnable.
View return policy
Description
SHPK Monoclonal antibody specifically detects SHPK in Human samples. It is validated for Immunocytochemistry, Western BlotSpecifications
| SHPK | |
| Monoclonal | |
| 0.63 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P50440 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 2628 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Immunocytochemistry, Western Blot | |
| OTI1D2 | |
| Unconjugated | |
| GATM | |
| 4930431K22Rik; AI194947; AW260459; Carbohydrate kinase-like protein; CARKL; Sedoheptulokinase; SHK; Shpk | |
| Full length human recombinant protein of human GATM produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |