missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SHPRH Monoclonal Antibody (OTI4A3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA501448S
This item is not returnable.
View return policy
Description
SHPRH Monoclonal antibody specifically detects SHPRH in Human samples. It is validated for Western BlotSpecifications
| SHPRH | |
| Monoclonal | |
| 0.88 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q149N8 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 257218 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Western Blot | |
| OTI4A3 | |
| Unconjugated | |
| SHPRH | |
| 2610103K11Rik; AA450458; AU024614; bA545I5.2; BC006883; D230017O13Rik; E130018M05; E3 ubiquitin-protein ligase SHPRH; KIAA2023; RING-type E3 ubiquitin transferase SHPRH; Shprh; SNF2 histone linker PHD RING helicase; SNF2 histone linker PHD RING helicase, E3 ubiquitin protein ligase; SNF2, histone-linker, PHD and RING finger domain-containing helicase | |
| Human recombinant protein fragment corresponding to amino acids 1-100 and 1500-1600 of human SHPRH produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction