Learn More
SHPRH Monoclonal Antibody (OTI5G8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA501447
Encompass_Preferred
 View more versions of this product
Product Code. 17544271
  • € 540.87 / 100µL
Description
A novel gene containing SNF2, PHD-finger, RING-finger, helicase, and linker histone domains termed SHPRH, is a possible candidate for a tumor suppressor gene. These domains are characteristic of several DNA repair proteins, transcription factors, and helicases. Mapping of this gene to 6q24, a region reported to contain a tumor suppressor locus suggests a role in tumor suppression.
Specifications
Specifications
SHPRH
Monoclonal
0.79 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q149N8
SHPRH
Human recombit protein fragment corresponding to amino acids 1-100 and 1500-1600 of human SHPRH produced in HEK293T cell.
100 μL
Primary
Human
Antibody
IgG2b
Flow Cytometry, Western Blot
OTI5G8
Unconjugated
SHPRH
bA545I5.2
Mouse
Affinity Chromatography
RUO
257218
-20° C, Avoid Freeze/Thaw Cycles