Learn More
SHPRH Mouse anti-Human, Clone: OTI5G8, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF501447
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
A novel gene containing SNF2, PHD-finger, RING-finger, helicase, and linker histone domains termed SHPRH, is a possible candidate for a tumor suppressor gene. These domains are characteristic of several DNA repair proteins, transcription factors, and helicases. Mapping of this gene to 6q24, a region reported to contain a tumor suppressor locus suggests a role in tumor suppression.Specifications
| SHPRH | |
| Monoclonal | |
| Unconjugated | |
| SHPRH | |
| bA545I5.2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 257218 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Flow Cytometry, Western Blot | |
| OTI5G8 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q149N8 | |
| SHPRH | |
| Human recombit protein fragment corresponding to amino acids 1-100 and 1500-1600 of human SHPRH produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.