missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SHROOM1 Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-82005B
This item is not returnable.
View return policy
Description
SHROOM1 Polyclonal antibody specifically detects SHROOM1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
Specifications
| SHROOM1 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| 134549 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, ELISA | |
| Biotin | |
| Apical Protein 2, APXL2, KIAA1960, Protein Shroom1, Shroom Family Member 1 | |
| Rabbit polyclonal SHROOM1 antibody was raised against a 16 amino acid peptide near the carboxy terminus of human SHROOM1. The immunogen is located within the last 50 amino acids of SHROOM1. Amino Acid Squence: CDDLGHHAPSPSPARPPGT | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction