missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SIAH1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16609084
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16609084 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16609084 Supplier Invitrogen™ Supplier No. PA5102615

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total SIAH1.

This gene encodes a protein that is a member of the seven in absentia homolog family. The protein is an E3 ligase and is involved in ubiquitination and proteasome-mediated degradation of specific proteins. The activity of this ubiquitin ligase has been implicated in the development of certain forms of Parkinson's disease, the regulation of the cellular response to hypoxia and induction of apoptosis. Alternative splicing results in several additional transcript variants, some encoding different isoforms and others that have not been fully characterized.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SIAH1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SIAH1
Gene Accession No. P61092, Q06985, Q8IUQ4, Q920M9
Gene Alias AA960570; AA982064; AI853500; E3 ubiquitin-protein ligase SIAH1; E3 ubiquitin-protein ligase SIAH1A; E3 ubiquitin-protein ligase SIAH1B; FLJ08065; h HUMSIAH; hSIAH1; HUMSIAH; mSiah-1a; RING-type E3 ubiquitin transferase SIAH1; RING-type E3 ubiquitin transferase SIAH1A; RING-type E3 ubiquitin transferase SIAH1B; seven in absentia 1A; seven in absentia 1B; seven in absentia homolog 1; Seven in absentia homolog 1a; Seven in absentia homolog 1b; SIAH; siah E3 ubiquitin protein ligase 1; SIAH1; siah-1; Siah1a; Siah-1a; Siah1b; siah-1b; Sinh1a; Sinh1b
Gene Symbols SIAH1, Siah1a, Siah1b
Host Species Rabbit
Immunogen A synthesized peptide derived from human SIAH1(Accession Q8IUQ4), corresponding to amino acid residues M1-N33.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 110337437, 140941, 20437, 20438, 6477
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.