missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SIAH2 Monoclonal Antibody (24E6H3)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA524780
This item is not returnable.
View return policy
Description
This is specific for human SIAH2 and Drosophila SINA in proliferating cells and stem cells. This does not detect Drosophila SINAH or normal, nondividing cells.
SIAH-2 (seven in absentia homolog 2) is an E3 ligase that catalyzes ubiquitination and proteasome-mediated degradation of protein substrates. SIAH-2 encodes a 324 amino acid protein that shares 77% identity with human SIAH-1 and 68% identity with the Drosophila sina (seven in absentia) gene, on which development of the Drosophila R7 photoreceptor is dependent. SIAH-2 targets TRAF2 (which regulates cell responses to stress and cytokines through the regulation of key stress-signaling cascades) for degradation under stress conditions such as hypoxia. It targets HIF-1 alpha prolyl hydroxylase 3 (PHD3) for degradation upon exposure to hypoxic conditions, which coincides with an increase in SIAH-2 transcription. SIAH-2 can decrease TNF-alpha-dependent induction of JNK activity and transcriptional activation of NFkB. SIAH-2 null mice subjected to hypoxia display an impaired respiratory response and reduced levels of hemoglobin.
Specifications
| SIAH2 | |
| Monoclonal | |
| Conc. Not Determined | |
| Tissue culture supernatant with 0.1% sodium azide | |
| O43255, P21461, Q06986 | |
| SIAH2, sina | |
| Synthetic peptide made to an N-terminal portion of the drosophila SINA protein (within residues 1-100). | |
| RUO | |
| 100622531, 20439, 39884, 6478 | |
| 4°C, do not freeze | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 24E6H3 | |
| Unconjugated | |
| SIAH2 | |
| AA415433; E3 ubiquitin-protein ligase SIAH2; EC 6.3.2; hSiah2; mSiah2; RING-type E3 ubiquitin transferase SIAH2; seven in absentia 2; seven in absentia homolog 2; siah E3 ubiquitin protein ligase 2; SIAH2; siah-2; Sinh2 | |
| Mouse | |
| 500 μL | |
| Primary | |
| Drosophila, Human, Mouse, Pig | |
| Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction