missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ SIB 1508Y maleate

Product Code. p-5546024
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18725047

Brand: Tocris Bioscience™ 4766/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent agonist of α4β2, α2β4, α4β4 and α3β4 nACh receptors

SIB 1508Y maleate is a potent neuronal nicotinic ACh receptor agonist (EC50 values are 1.8, 5, 9 and 23 nM for α4β2, α2β4, α4β4 and α3β4 receptors respectively). Improves cognitive function in a MPTP-induced model of Parkinson's Disease in vivo.
TRUSTED_SUSTAINABILITY

Specifications

Purity >98%
Quantity 10 mg
Product Type SIB 1508Y maleate
Molecular Weight (g/mol) 302.33
Inhibitors SIB 1508Y maleate

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.