missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SIGB Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5144540
This item is not returnable.
View return policy
Description
SIGB Polyclonal Antibody for Western Blot, ELISA
Required for the transition of plastids into chloroplasts by coordinating nuclear and chloroplastic genomes under light conditions. Sigma factors are initiation factors that promote the attachment of plastid-encoded RNA polymerase (PEP) to specific initiation sites and are then released. Promotes the biosynthesis of plastid-encoded tRNAs (e.g. trnE-UUC and trnV-UAC).
Specifications
| SIGB | |
| Polyclonal | |
| Unconjugated | |
| SIGB | |
| ABC1; SIG1; SIG2; SIGA | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 837376 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot | |
| 2.274 mg/mL | |
| PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
| O22056 | |
| SIGB | |
| Recombinant Arabidopsis thaliana SIGB protein (40-310aa). | |
| 100 μL | |
| Primary | |
| Plant | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction