missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SIGIRR Recombinant Rabbit Monoclonal Antibody (HL2008)
GREENER_CHOICE

Product Code. 18095551
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18095551 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18095551 Supplier Invitrogen™ Supplier No. MA550360

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

SIGIRR Recombinant Monoclonal Antibody for Western Blot, IHC (P)

SIGIRR (single Ig IL-1R-related molecule) is a novel member of the interleukin 1 receptor (IL-1R) superfamily. Although it shows the typical conserved motifs that characterize the IL-1R and Toll superfamily, it is structurally and functionally distinct from both. SIGIRR has only one Ig domain in its extracellular portion whereas the IL-1R family contains three Ig folds. An unusually long cytoplasmic domain is reminiscent of the structure of drosophila Toll, yet the SIGIRR peptide sequence is more closely related to IL-1RI. A chimera, in which the SIGIRR intracellular domain was fused to the IL-1R extracellular domain, did not activate NFkappaB unlike similar fusion proteins of other IL-1R related molecules. Phylogenetic data that suggest that the plasma membrane protein SIGIRR is a close functional relative of MyD88 in mammals.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SIGIRR
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone HL2008
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene SIGIRR
Gene Accession No. Q4V892, Q6IA17, Q9JLZ8
Gene Alias AI256711; IL-1R8; MGC110992; Sigirr; single Ig and TIR domain containing; single Ig IL-1 receptor related protein; single Ig IL-1-related receptor; Single Ig IL-1R-related molecule; single Ig IL-1R-related protein; single immunoglobulin and toll-interleukin 1 receptor (TIR) domain; single immunoglobulin domain IL1R1 related; single immunoglobulin domain-containing IL1R-related protein; TIR8; Toll/interleukin-1 receptor 8; UNQ301/PRO342
Gene Symbols SIGIRR
Host Species Rabbit
Immunogen Recombinant fragment of human SIGIRR.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24058, 309106, 59307
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.