missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Siglec-1/CD169 Antibody (3D6.112 (recombinant version)) - Azide and BSA Free, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Siglec-1/CD169 Monoclonal antibody specifically detects Siglec-1/CD169 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Specifications
Specifications
| Antigen | Siglec-1/CD169 |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Clone | 3D6.112 (recombinant version) |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS |
| Gene Alias | CD169, FLJ00051, sialic acid binding Ig-like lectin 1, sialoadhesin, sialoadhesin |
| Host Species | Rat |
| Immunogen | Spleen cells from AO rats immunised with thioglycollate-elicted murine peritoneal macrophages (TPM) that had been cutured in mouse serum to induce sheep erythrocyte receptor (SER). |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?