missing translation for 'onlineSavingsMsg'
Learn More

Siglec-1/CD169 Antibody (3D6.112 (recombinant version)) - Azide and BSA Free, Novus Biologicals™

Product Code. p-200072408 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
Unit Size:
0.1mg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18657006 0.025 mg 25µg
18650069 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18657006 Supplier Novus Biologicals Supplier No. NBP3089900.025mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

Siglec-1/CD169 Monoclonal antibody specifically detects Siglec-1/CD169 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Siglec-1/CD169
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone 3D6.112 (recombinant version)
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS
Gene Alias CD169, FLJ00051, sialic acid binding Ig-like lectin 1, sialoadhesin, sialoadhesin
Host Species Rat
Immunogen Spleen cells from AO rats immunised with thioglycollate-elicted murine peritoneal macrophages (TPM) that had been cutured in mouse serum to induce sheep erythrocyte receptor (SER).
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 6614
Target Species Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.