missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Siglec-1/CD169 Antibody (645608), PE/Cy5.5, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB5610PECY55
This item is not returnable.
View return policy
Description
Siglec-1/CD169 Monoclonal antibody specifically detects Siglec-1/CD169 in Mouse samples. It is validated for Flow Cytometry
Specifications
| Siglec-1/CD169 | |
| Monoclonal | |
| PE-Cyanine5.5 | |
| PBS | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry | |
| 645608 | |
| Flow Cytometry | |
| CD169, FLJ00051, sialic acid binding Ig-like lectin 1, sialoadhesin, sialoadhesin | |
| Mouse myeloma cell line NS0-derived recombinant mouse Siglec-1/CD169, Thr20-Leu1639 (predicted), Accession # Q62230 | |
| 0.1 mL | |
| Adaptive Immunity, Immunology | |
| 6614 | |
| Store at 4C in the dark. Do not freeze. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction