missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Siglec-1/CD169 Antibody (7-239), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-37729AF750
This item is not returnable.
View return policy
Description
Siglec-1/CD169 Monoclonal specifically detects Siglec-1/CD169 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications
| Siglec-1/CD169 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| SIGLEC1 | |
| Human rhinovirus 14-infected monocyte-derived dendritic cells. | |
| Primary | |
| The mouse monoclonal antibody 7-239 recognizes CD169 (sialoadhesin, Siglec-1), a 210 kDa type I transmembrane glycoprotein expressed on macrophages and dendritic cells. | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) | |
| 7-239 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen | |
| CD169, FLJ00051, sialic acid binding Ig-like lectin 1, sialoadhesin, sialoadhesin | |
| Mouse | |
| 0.1 mL | |
| 6614 | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction