missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Siglec-1/CD169 Antibody (HSn 7D2), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08615UV
This item is not returnable.
View return policy
Description
Siglec-1/CD169 Monoclonal antibody specifically detects Siglec-1/CD169 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| Siglec-1/CD169 | |
| Monoclonal | |
| DyLight 350 | |
| CD169, FLJ00051, sialic acid binding Ig-like lectin 1, sialoadhesin, sialoadhesin | |
| Fc fusion protein containing N-terminal 4 domains of human Siglec-1/CD169. (Uniprot: Q9ZZ2) | |
| 100 μg | |
| Adaptive Immunity, Immunology | |
| 6614 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| HSn 7D2 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction