missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Siglec-10 Antibody (5G6) [Janelia Fluor™ 635], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-21494JF635
This item is not returnable.
View return policy
Description
Siglec-10 Monoclonal antibody specifically detects Siglec-10 in Human samples. It is validated for ELISA,Immunohistochemistry,Immunoprecipitation
Specifications
| Siglec-10 | |
| Monoclonal | |
| Janelia Fluor 635 | |
| MGC126774, PRO940, sialic acid binding Ig-like lectin 10, sialic acid-binding Ig-like lectin 10, SIGLEC-10, siglec-like gene 2, Siglec-like protein 2, SLG2sialic acid binding Ig-like lectin 10 Ig-like lectin 7 | |
| Siglec-10 extracellular domain fused with human IgG1 Fc fragment | |
| 0.1 mL | |
| Immunology | |
| 89790 | |
| Store at 4°C in the dark. | |
| IgG1 |
| ELISA, Immunohistochemistry, Immunoprecipitation | |
| 5G6 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction