Learn More
Siglec-10 Antibody (5G6), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-21494PCP
Product Code. 18615344
  • € 577.00 / 0.10mL
Description
Siglec-10 Monoclonal antibody specifically detects Siglec-10 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunoprecipitation
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ELISA, Immunohistochemistry, Immunoprecipitation
PBS
Store at 4C in the dark.