missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Siglec-10 Antibody (SIGLEC10/7582), Novus Biologicals™

Mouse Monoclonal Antibody

€ 232.00 - € 462.00

Specifications

Antigen Siglec-10
Clone SIGLEC10/7582
Concentration 0.2 mg/mL
Dilution Immunohistochemistry-Paraffin 1-2 μg/mL for 30 minutes at RT
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30043271
View Documents
Novus Biologicals™
NBP3-26756-100ug
100 μg
€ 462.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30043823
View Documents
Novus Biologicals™
NBP3-26756-20ug
20 μg
€ 232.00
20µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Siglec-10 Monoclonal antibody specifically detects Siglec-10 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

Siglec-10
0.2 mg/mL
Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Immunology
10 mM PBS with 0.05% BSA
89790
IgG2 κ
Protein A or G purified
SIGLEC10/7582
Immunohistochemistry-Paraffin 1-2 μg/mL for 30 minutes at RT
Monoclonal
Purified
RUO
Human
MGC126774, PRO940, sialic acid binding Ig-like lectin 10, sialic acid-binding Ig-like lectin 10, SIGLEC-10, siglec-like gene 2, Siglec-like protein 2, SLG2sialic acid binding Ig-like lectin 10 Ig-like lectin 7
Recombinant fragment (around aa500-697) of human Siglec-10 protein (exact sequence is proprietary)
Primary
Store at 4°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.