missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Siglec-11 Antibody (705904) [DyLight 755], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB3258Z
This item is not returnable.
View return policy
Description
Siglec-11 Monoclonal antibody specifically detects Siglec-11 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOF
Specifications
| Siglec-11 | |
| Monoclonal | |
| DyLight 755 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunohistochemistry, CyTOF | |
| 705904 | |
| Flow Cytometry, Immunohistochemistry, CyTOF-ready | |
| sialic acid binding Ig-like lectin 11, sialic acid-binding Ig-like lectin 11, Sialic acid-binding lectin 11, siglec-11 | |
| Human Siglec-11 synthetic peptide, PPARLSWTRWGQTVGPSQPSDPGVLELPPIC, Accession # Q96RL6 | |
| 0.1 mL | |
| Immunology | |
| 114132 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction