missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Siglec-14 Antibody (2457E), DyLight 550, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB4905L
This item is not returnable.
View return policy
Description
Siglec-14 Monoclonal antibody specifically detects Siglec-14 in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
| Siglec-14 | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 100049587 | |
| Store at 4°C in the dark. | |
| IgG |
| Flow Cytometry, CyTOF | |
| 2457E | |
| Flow Cytometry, CyTOF-ready | |
| AC018755.2, sialic acid binding Ig-like lectin 14 | |
| Mouse myeloma cell line, NS0-derived human Siglec-14, Lys18-Leu358, Accession # Q08ET2 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction