missing translation for 'onlineSavingsMsg'
Learn More

Siglec-14 Antibody (2457E) [Janelia Fluor™ 635], Novus Biologicals™

Product Code. 30168615 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30168615 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30168615 Supplier Novus Biologicals Supplier No. FAB4905JF635

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Siglec-14 Monoclonal antibody specifically detects Siglec-14 in Human samples. It is validated for Flow Cytometry,CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen Siglec-14
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 2457E
Conjugate Janelia Fluor 635
Formulation 50mM Sodium Borate
Gene Alias AC018755.2, sialic acid binding Ig-like lectin 14
Host Species Rabbit
Immunogen Mouse myeloma cell line, NS0-derived human Siglec-14, Lys18-Leu358, Accession # Q08ET2
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100049587
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.