missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Siglec-15 Antibody (3F7F3), Novus Biologicals™
Mouse Monoclonal Antibody
€ 298.00 - € 511.00
Specifications
| Antigen | Siglec-15 |
|---|---|
| Clone | 3F7F3 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/ Immunofluorescence 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 |
| Applications | Flow Cytometry, ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30029529
|
Novus Biologicals™
NBP3-27165-0.1ml |
0.1 mL |
€ 511.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30025499
|
Novus Biologicals™
NBP3-27165-0.025ml |
0.025 mL |
€ 298.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Siglec-15 Monoclonal antibody specifically detects Siglec-15 in Human,Monkey samples. It is validated for Flow Cytometry,ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ ImmunofluorescenceSpecifications
| Siglec-15 | |
| 1 mg/mL | |
| Flow Cytometry, ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Immunology | |
| PBS | |
| 284266 | |
| IgG1 | |
| Protein G purified |
| 3F7F3 | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/ Immunofluorescence 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Monkey | |
| CD33 antigen-like 3SIGLEC-15, CD33 molecule-like 3, CD33L3, HsT1361, sialic acid binding Ig-like lectin 15, sialic acid-binding Ig-like lectin 15, Siglec-15 | |
| Purified recombinant fragment of human Siglec-15 (AA: Extra(20-263)) expressed in Mammal. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title