missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Siglec-15 Antibody (AB-25E9), PE, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals NBP3-28008PE
This item is not returnable.
View return policy
Description
Siglec-15 Monoclonal antibody specifically detects Siglec-15 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| Siglec-15 | |
| Monoclonal | |
| PE | |
| CD33 antigen-like 3SIGLEC-15, CD33 molecule-like 3, CD33L3, HsT1361, sialic acid binding Ig-like lectin 15, sialic acid-binding Ig-like lectin 15, Siglec-15 | |
| Siglec-15/ CD33L3 | |
| 0.1 mL | |
| Immunology | |
| 284266 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| AB-25E9 | |
| PBS | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction