missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Siglec-16 Antibody (706030), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB68192D
This item is not returnable.
View return policy
Description
Siglec-16 Monoclonal antibody specifically detects Siglec-16 in Human samples. It is validated for Flow Cytometry, Flow Cytometry (Intracellular Staining), CyTOF
Specifications
| Siglec-16 | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 400709 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, Flow Cytometry (Intracellular Staining), CyTOF | |
| 706030 | |
| Flow Cytometry, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| Siglec-P16, SIGLECP16 | |
| Human Siglec-16 peptide, Accession # A6NMB1 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction