missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Siglec-16 Antibody (706045) [Janelia Fluor™ 635], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB68191JF635
This item is not returnable.
View return policy
Description
Siglec-16 Monoclonal antibody specifically detects Siglec-16 in Human samples. It is validated for Flow Cytometry,Flow Cytometry (Intracellular Staining),CyTOF
Specifications
| Siglec-16 | |
| Monoclonal | |
| Janelia Fluor 635 | |
| Siglec-P16, SIGLECP16 | |
| Human Siglec-16 peptide, Accession # A6NMB1 | |
| RUO | |
| 400709 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Flow Cytometry, Flow Cytometry (Intracellular Staining), CyTOF | |
| 706045 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction