missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Siglec-2/CD22 Antibody (OTI1F12), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-46125
This item is not returnable.
View return policy
Description
Ensure accurate, reproducible results in Western Blot, Immunofluorescence
Siglec-2/CD22 Monoclonal specifically detects Siglec-2/CD22 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| Siglec-2/CD22 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:400 - 1:4000, Immunocytochemistry/Immunofluorescence | |
| B-cell receptor CD22, BL-CAM, B-lymphocyte cell adhesion molecule, CD22 antigenMGC130020, CD22 molecule, sialic acid binding Ig-like lectin 2, Sialic acid-binding Ig-like lectin 2, SIGLEC-2, SIGLEC2FLJ22814, T-cell surface antigen Leu-14 | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey | |
| Purified |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| OTI1F12 | |
| Unconjugated | |
| PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
| CD22 | |
| Full length human recombinant protein of human CD22(NP_001762) produced in HEK293T cell. | |
| 0.1 mL | |
| Adaptive Immunity, DNA Repair, Immunology | |
| 933 | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction