missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Siglec-3/CD33 Antibody (CD33 3D6), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50291G
This item is not returnable.
View return policy
Description
Siglec-3/CD33 Monoclonal specifically detects Siglec-3/CD33 in Human samples. It is validated for ELISA.
Specifications
| Siglec-3/CD33 | |
| Monoclonal | |
| DyLight 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| CD33 | |
| Siglec-3/CD33-Fc recombinant protein | |
| 0.1 mL | |
| Immunology | |
| 945 | |
| Store at 4C in the dark. | |
| IgG1 |
| ELISA | |
| CD33 3D6 | |
| ELISA | |
| CD33 antigen, CD33 antigen (gp67), CD33 molecule, FLJ00391, myeloid cell surface antigen CD33, p67, sialic acid binding Ig-like lectin 3, Sialic acid-binding Ig-like lectin 3, SIGLEC-3, SIGLEC3gp67 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction