missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Siglec-3/CD33 Antibody (domagrozumab), Alexa Fluor™ 532, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals NBP3-28042AF532
This item is not returnable.
View return policy
Description
Siglec-3/CD33 Monoclonal antibody specifically detects Siglec-3/CD33 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| Siglec-3/CD33 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| CD33 antigen, CD33 antigen (gp67), CD33 molecule, FLJ00391, myeloid cell surface antigen CD33, p67, sialic acid binding Ig-like lectin 3, Sialic acid-binding Ig-like lectin 3, SIGLEC-3, SIGLEC3gp67 | |
| Siglec-3/ CD33 | |
| 0.1 mL | |
| Immunology | |
| 945 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| domagrozumab | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction