Learn More
Siglec-3/CD33 Antibody (Gemtuzumab), CoraFluor™ 1, Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals NBP3-28474CL1
Product Code. 30155580
  • € 844.00 / 0.10mL
Description
Siglec-3/CD33 Monoclonal antibody specifically detects Siglec-3/CD33 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Siglec-3/CD33
Monoclonal
CoraFluor 1
CD33 antigen, CD33 antigen (gp67), CD33 molecule, FLJ00391, myeloid cell surface antigen CD33, p67, sialic acid binding Ig-like lectin 3, Sialic acid-binding Ig-like lectin 3, SIGLEC-3, SIGLEC3gp67
Siglec-3/ CD33
0.1 mL
Immunology
945
Store at 4°C in the dark. Do not freeze.
IgG4
Flow Cytometry, ELISA, Functional Assay
Gemtuzumab
PBS
Human
Protein A purified