missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Siglec-5/CD170 Antibody (110), Janelia Fluor™ 549, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89986JF549
This item is not returnable.
View return policy
Description
Siglec-5/CD170 Monoclonal specifically detects Siglec-5/CD170 in Human samples. It is validated for Flow Cytometry.
Specifications
| Siglec-5/CD170 | |
| Monoclonal | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 110 | |
| Flow Cytometry | |
| CD170, CD170 antigen, CD33 antigen-like 2, CD33L2OB binding protein-2, OB-BP2OB-binding protein 2, OBBP2sialic acid-binding Ig-like lectin 5, Obesity-binding protein 2, sialic acid binding Ig-like lectin 5, sialic acid-binding immunoglobulin-like lectin 5, SIGLEC-5 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Siglec-5/CD170 (O15389; Met1-Thr 434). | |
| 0.1 mL | |
| Immunology | |
| 8778 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction