missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Siglec-5/Siglec-14 Antibody (194128), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB10721C
This item is not returnable.
View return policy
Description
Siglec-5/Siglec-14 Monoclonal antibody specifically detects Siglec-5/Siglec-14 in Human samples. It is validated for Flow Cytometry, ELISA Standard (Matched Pair), Neutralization, ELISA (Matched Antibody Pair), ELISA (Matched Antibody Pair), CyTOF
Specifications
| Siglec-5/Siglec-14 | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 8778 | |
| Store at 4C in the dark. | |
| IgG1 |
| Flow Cytometry, ELISA Standard (Matched Pair), Neutralization, ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair) | |
| 194128 | |
| Flow Cytometry, ELISA Standard (Matched Pair), Neutralization, ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), CyTOF-ready | |
| CD170,CD33L2,OBBP2,OB-BP2,Sialic acid-binding Ig-like lectin 5,SIGLEC5,SIGLEC-5 | |
| Mouse myeloma cell line NS0-derived recombinant human Siglec-5/Siglec-14, Lys18-Thr434, Accession # O15389 | |
| 0.5 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction