missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Siglec-7/CD328 Antibody (194212), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB1138C
This item is not returnable.
View return policy
Description
Siglec-7/CD328 Monoclonal antibody specifically detects Siglec-7/CD328 in Human samples. It is validated for Western Blot
Specifications
| Siglec-7/CD328 | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| 194212 | |
| Western Blot | |
| Adhesion inhibitory receptor molecule 1, adhesion inhibitory receptor molecule 1, siglec-7, AIRM-1, AIRM1QA79 membrane protein, CD328, CD328 antigen, CDw328, D-siglec, p75, p75/AIRM1, QA79, sialic acid binding Ig-like lectin 7, sialic acid binding immunoglobulin-like lectin 7, sialic acid-binding Ig-like lectin 7, Siglec-7 | |
| Mouse myeloma cell line NS0-derived recombinant human Siglec-7/CD328, Gln19-Gly357 (predicted), Accession # Q9Y286 | |
| 0.5 mL | |
| Cardiovascular Biology | |
| 27036 | |
| Store at 4C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction