Learn More
Siglec-E Antibody (750620) [PE], Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB5806P
This item is not returnable.
View return policy
Description
Siglec-E Monoclonal antibody specifically detects Siglec-E in Mouse samples. It is validated for Flow Cytometry, CyTOF
Specifications
| Siglec-E | |
| Monoclonal | |
| PE | |
| PBS | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 8778 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Flow Cytometry, CyTOF | |
| 750620 | |
| Flow Cytometry, CyTOF-ready | |
| CD170;CD33L2;OBBP2;OB-BP2;Sialic acid-binding Ig-like lectin 5;SIGLEC5;SIGLEC-5 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Siglec-E, Gln19-Phe354, Accession # Q91Y57 | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Purified |