Learn More
Siglec-E Antibody (750620) [PE], Novus Biologicals™
Rat Monoclonal Antibody
Brand:  Novus Biologicals FAB5806P
Product Code. 18686144
  • € 689.00 / 0.10mL
Description
Siglec-E Monoclonal antibody specifically detects Siglec-E in Mouse samples. It is validated for Flow Cytometry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Siglec-E
Monoclonal
PE
PBS
Rat
Protein A or G purified from hybridoma culture supernatant
RUO
8778
Store at 4°C in the dark.
IgG2b
Flow Cytometry, CyTOF
750620
Flow Cytometry, CyTOF-ready
CD170;CD33L2;OBBP2;OB-BP2;Sialic acid-binding Ig-like lectin 5;SIGLEC5;SIGLEC-5
Mouse myeloma cell line NS0-derived recombinant mouse Siglec-E, Gln19-Phe354, Accession # Q91Y57
0.1 mL