Learn More
Invitrogen™ SIGLEC10 Monoclonal Antibody (5G6)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA528237
Description
Mouse anti Human Siglec-10 antibody, clone 5G6 recognizes human Siglec-10 (Sialic acid-binding Ig-like lectin 10), a putative adhesion molecule and member of the Ig superfamily, expressed by monocytes, B cells, eosinophils, and at a higher level by a subpopulation of CD16+CD56- natural killer (NK) cells.
SIGLECs are members of the immunoglobulin superfamily that are expressed on the cell surface. Most SIGLECs have 1 or more cytoplasmic immune receptor tyrosine-based inhibitory motifs, or ITIMs. SIGLECs are typically expressed on cells of the innate immune system, with the exception of the B-cell expressed SIGLEC6.
Specifications
| SIGLEC10 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.09% sodium azide | |
| Q96LC7 | |
| SIGLEC10 | |
| Recombinant human Siglec-10, fused with the Fc region of human IgG. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry | |
| 5G6 | |
| Unconjugated | |
| SIGLEC10 | |
| PRO940; sialic acid binding Ig like lectin 10; sialic acid binding Ig-like lectin 10; sialic acid binding Ig-like lectin 10 Ig-like lectin 7; sialic acid binding Ig-like lectin G; sialic acid-binding Ig-like lectin 10; Siglec; SIGLEC10; Siglec-10; Siglecg; siglec-like gene 2; siglec-like protein 2; SLG2; UNQ477/PRO940 | |
| Mouse | |
| Protein A | |
| RUO | |
| 89790 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.