missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Sigma 54 Monoclonal Antibody (33.7)
GREENER_CHOICE

Mouse Monoclonal Antibody

€ 510.00 - € 624.00

Specifications

Antigen Sigma 54
Clone 33.7
Concentration 1 mg/mL
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Applications ELISA, Western Blot
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11544383
GREENER_CHOICE
View Documents
Invitrogen™
MA15876
100 μL
€ 624.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11554383
View Documents
Invitrogen™
MA15877
50 μL
€ 510.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This antibody binds specifically in ELISA to monomeric tetrameric Sigma 54 but it does not bind to RAT protein or AgrA protein.

Sigma factors are initiation factors that promote the attachment of RNA polymerase to specific initiation sites and are then released. This sigma factor is responsible for the expression of enzymes involved in arginine catabolism. The open complex (sigma54 and core RNA polymerase) serves as the receptor for the receipt of the melting signal from the remotely bound activator protein glnG(ntrC).
Specifications

Specifications

Sigma 54
1 mg/mL
ELISA, Western Blot
Unconjugated
Mouse
RUO
PBS with 0.1% sodium azide
Primary
Protein G
33.7
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Monoclonal
Liquid
IgG1 κ
Bacteria
Bacterially expressed Sigma 54 from Balb/c Sp2/0 cells, purified over a heparin column.
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.