missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SIGNR1/CD209b Antibody (123), HRP , Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90608H
This item is not returnable.
View return policy
Description
SIGNR1/CD209b Monoclonal antibody specifically detects SIGNR1/CD209b in Mouse samples. It is validated for ELISA, ELISA
Specifications
| SIGNR1/CD209b | |
| Monoclonal | |
| HRP | |
| CD209 antigen like protein B, DC SIGN related protein 1, DC SIGNR1, OtB7 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Mouse SIGNR1/CD209b (NP_081248.2; Gln74-Gly325). | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Purified |
| ELISA | |
| 123 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 170786 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction