missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SIGNR4/CD209e Antibody (RM0327-9B29), Alexa Fluor™ 750, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-12431AF750
This item is not returnable.
View return policy
Description
SIGNR4/CD209e Monoclonal specifically detects SIGNR4/CD209e in Mouse samples. It is validated for Western Blot.
Specifications
| SIGNR4/CD209e | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| Mouse recombinant SIGNR4/CD209e | |
| 0.1 mL | |
| Mouse | |
| IgG2 |
| Western Blot | |
| RM0327-9B29 | |
| Western Blot | |
| Rat | |
| Protein A or G purified | |
| Primary | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction