missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SIK2 Mouse anti-Human, Clone: S15G10, Abnova™
Mouse monoclonal antibody raised against recombinant human SIK2.
Brand: Abnova MAB12947.50ug
This item is not returnable.
View return policy
Specifications
| SIK2 | |
| Monoclonal | |
| Unconjugated | |
| Immunocytochemistry Immunofluorescence (1:100-1:400) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) Immunoprecipitation Western Blot (1:250) The optimal working dilution should be determined by the end user. | |
| DKFZp434K1115/KIAA0781/LOH11CR1I/QIK/SNF1LK2 | |
| Mouse | |
| Affinity purification | |
| RUO | |
| 23235 | |
| Store at 4°C. Do not freeze. | |
| IgG1 |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot | |
| S15G10 | |
| Mouse monoclonal antibody raised against recombinant human SIK2. | |
| In PBS, pH 7.2 (0.09% sodium azide). | |
| SIK2 | |
| Recombinant protein corresponding to human SIK2. | |
| 50 μg | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction