Learn More
Cole-Parmer™ Silastic Laboratory Tubing
Brand:  Cole-Parmer™ 96115-08
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11759989
  • € 81.06 / Each
please call 020 4877000
Add to basket
Specifications
USP,BP,GMP,FDA, ISO–9002
Autoclavable
Rubber
0.6 mm
-54°C
249°C
1 mm
2.2 mm
For Pharmaceutical, Biotech, Filtration, and Environmental Analysis
Sterile