missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Silastic™ Laboratory Tubing

Ideal for pharmaceutical, biotech, filtration, and environmental analysis applications

Brand:  Cole-Parmer™ 96115-10

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 11736018

  • € 75.37 / Each


please call 020 4877000
Add to basket

Description

Description

  • Platinum-cured, nonpyrogenic tubing.
  • Bacteria and fungus-resistant.
  • Extremely low extractables.
  • Does not impart a taste or odor.
  • Exceeds USP Class VI Plastics Test requirements.
  • Elastomer and tubing are manufactured under Bulk Pharmaceutical cGMPs in an FDA-compliant, ISO-9002-certified facility.
  • Sterilize: Autoclave or radiation.
Specifications

Specifications

3A;ADCF;FDA;European Pharmacopoeia;Non-DEHP;Non-Phthalate;NSF-61;NSF-51;USP
Platinum-cured, nonpyrogenic, extremely low extractables, and does not impart a taste or odor
1.5 mm
50 ft.
0.24 mm
-65°F
50 ft./Pk.
Autoclave or radiation
248°F
Autoclavable
2.0 mm
15.2 m
Pharmaceutical, biotech, filtration, and environmental analysis.
-53°C
Bacteria and fungus-resistant.
Videos
SDS
Documents