Learn More
Sildenafil-d8 (piperazine-d8), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Sildenafil-D8 |
| CAS | 951385-68-5 |
| Molecular Formula | C22 2H8 H22 N6 O4 S |
| Quantity | 0.01 g |
| Synonym | 7 H-Pyrazolo[4,3-d]pyrimidin-7-one, 5-[2-ethoxy-5-[(4-methyl-1-piperazinyl-2,2,3,3,5,5,6,6-d8), BDD 10402 sulfonyl]phenyl]-1,6-dihydro-1-methyl-3-propyl- |
| InChI Formula | InChI=1 S/C22H30N6O4S/c1-5-7-17-19-20(27(4)25-17)22(29)24-21(23-19)16-14-15(8-9-18(16)32-6-2)33(30,31)28-12-10-26(3)11-13-28/h8-9,14 H,5-7,10-13H2,1-4H3,(H,23,24,29)/i10D2,11D2,12D2,13D2 |
| SMILES | [2 H]C1([2 H])N(C)C([2 H])([2 H])C([2 H])([2 H])N(C1([2 H])[2 H])S(=O)(=O)c2ccc(OCC)c(c2)C3=Nc4c(CCC)nn(C)c4C(=O)N3 |
| IUPAC Name | 5-[2-ethoxy-5-(2,2,3,3,5,5,6,6-octadeuterio-4-methylpiperazin-1-yl)sulfonylphenyl]-1-methyl-3-propyl-6 H-pyrazolo[4,3-d]pyrimidin-7-one |
| Molecular Weight (g/mol) | 482.63 |
| Formula Weight | 482.2551 g/mol |
| Show More |