Learn More
Ambion™ Silencer™ Select Pre-Designed siRNA
Silencer™ Select Pre-Designed siRNA
€ 752.00 - € 2350.00
Specifications
Purity HPLC
View More Specs
Products 3
Product Code Brand Description Quantity Price Quantity & Availability  
Product Code Brand Description Quantity Price Quantity & Availability  
10157534
View Documents Promotion Details
Ambion™
4390821
Silencer™ Select Validated siRNA, 20nmol, HPLC Purity, –20°C Storage, Potent, Most Specific, Reliable, Guaranteed, Incorporate the Latest Improvements in siRNA design, Off-Target Effect Prediction Algorithms and Chemistry 20 nmol
€ 752.00
Each
View alternative products
Save Up to 
»
Estimated Shipment: 07-05-2024
to see stock.
10299774
View Documents Promotion Details
Ambion™
4390822
Silencer™ Select Validated siRNA, 40nmol, HPLC Purity, –20°C Storage, Potent, Most Specific, Reliable, Guaranteed, Incorporate the Latest Improvements in siRNA design, Off-Target Effect Prediction Algorithms and Chemistry 40 nmol
€ 828.00
Each
View alternative products
Save Up to 
»
Estimated Shipment: 07-05-2024
to see stock.
10574835
View Documents Promotion Details
Ambion™
4407270
Silencer™ Select Validated, 250nmol, HPLC Purity, –20°C Storage, Classic 21-mers which Incorporate the Latest Improvements in siRNA Design, Off-target Effect Prediction Algorithms and Chemistry, Silencer Select Pre-Designed siRNAs are Guaranteed-To-Silence Based on their Proven Design 250 nmol
€ 2350.00
Each
View alternative products
Save Up to 
»
Estimated Shipment: 07-05-2024
to see stock.
Description
Description
The Silencer™ Select siRNAs are classic 21-mers which incorporate the latest improvements in siRNA design, off-target effect prediction algorithms, and chemistry. A choice of Silencer™ Select siRNAs are available—Pre-Designed or Validated. Silencer™ Select Pre-Designed siRNAs are guaranteed-to-silence based on their proven design while the Silencer™ Select Validated siRNAs are functionally tested to reduce target gene expression. Sets of Silencer™ Select siRNA Libraries are also available in tubes or plate for high throughput screening. The Silencer™ Select Positive and Negative Control RNAs should be included in every siRNA experiment.
The benefits of using Silencer™ Select siRNAs include:
  • Potent—up to 100-fold more potent than currently available siRNAs and fewer off-target effects
  • Most Specific—LNA™ chemical modifications reduce off-target effects by up to 90%
  • Reliable—demonstrated improvements in consistency and reliability of phenotypic results
  • Guaranteed—100% guaranteed to silence, the best guarantee in the industry
  • Note: Silencer™ Pre-Designed & Validated siRNAs are designed using our first-generation algorithm and DO NOT include any chemical modifications. They are recommended for completing in vitro RNAi studies that were started using Silencer™ siRNAs. For new studies we recommend Silencer Select siRNAs.
    Approximate Turnaround Times*
  • Standard purification: 4 business days
  • HPLC purification: 6 business days (20 and 40 nmol); 15 business days (250 nmol)
  • In Vivo Ready: 15 business days (250 nmol)
  • Specifications
    Specifications
    HPLC