Learn More
Thermo Scientific™ Silica gel, C4-RP, 8%C, ca. 1,5 mmol/g, part. size 40-63μm
Shop All Thermo Scientific Chemicals Products
Click to view available options
:
50g
Specifications
Specifications
| Particle Size | 40 to 63 μm |
| Packaging | Plastic Bottle |
| Quantity | 50 g |
| Material | Silica Gel |